1x1 image pixel for data collection
GC Tool Kit for Agilent Instruments
GC Tool Kit for Agilent Instruments
GC Tool Kit for Agilent Instruments
GC Tool Kit for Agilent Instruments
GC Tool Kit for Agilent Instruments
GC Tool Kit for Agilent Instruments
GC Tool Kit for Agilent Instruments
Thermo Scientific™
GC Tool Kit for Agilent Instruments
Thermo Scientific™ GC Tool Kit for Agilent Instruments includes common consumable supplies and tools.
Have Questions?
Catalog NumberFor Use With (Equipment)
60180-786Agilent GC-MS Instruments
Catalog number 60180-786
Price (USD)
926.74
Each
Add to cart
For Use With (Equipment):
Agilent GC-MS Instruments
Price (USD)
926.74
Each
Add to cart
Thermo Scientific™ GC Tool Kit for Agilent Instruments includes common consumable supplies and tools.
  • Replacements included perform like original instrument parts
  • Parts list included for easy reordering
Includes:
Capillary installation gauge, injector wrench, septum nut removal tool, open-end wrenches in 7/16 x 1/2in. and 1/2 x 9/16in. sizes.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
DescriptionGC Tool Kit for Agilent Instruments
For Use With (Application)GC
For Use With (Equipment)Agilent GC-MS Instruments
TypeCapillary Tool Kit
Unit SizeEach
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link