Search Thermo Fisher Scientific
Save time and minimize the risk of cross-contamination with convenient 96-Well Plate Kits packaged with pre-assembled glass or PTFE vials.
Catalog Number | Description | Profile | Total Volume (Metric) Well | Material |
---|---|---|---|---|
60180-K104 | 96 Well Microplate with Sealing Mat and Cutting Tool | Round | 1100 μL | Silicone/PTFE |
60180-K100 | 96 Well Small Volume Microplate with Sealing Mat | Taper | 500 μL | Silicone/PTFE |
60180-K101 | 96 Well Small Volume Microplate with Sealing Mat and Cutting Tool | Taper | 700 μL | Silicone/PTFE |
60180-K106 | 96 Deep Well Plate with Sealing Mat | Round | 1500 μL | Silicone/PTFE |
60180-K107 | 96 Square Well Plate with Sealing Mat | Constricted | 1000 μL | Silicone/PTFE |
Save time and minimize the risk of cross-contamination with Thermo Scientific™ WebSeal™ 96-Well Plate Kits with Sealing Mats. Compatible with all 96-well-configured autosamplers, the ready-to-use kits consist of a comprehensive choice of 96-well microplates, pre-inserted glass or PTFE vials and silicone/PTFE sealing mats.