Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Effectively seal all microplate formats with Thermo Scientific™ WebSeal™ Sealing Tapes. Available in a selection of thin, lightweight materials and adhesives to meet the needs of any application or assay, they minimize evaporation and protect samples from contamination and spilling.
Color | Clear |
Description | Sealing Tape with SRA (synthetic rubber adhesive) |
Material | Polyolefin, Rubber |
No. per Pack | 100 |
Product Type | Non-sterile Sealing Tape |
Product Line | Webseal |
Adhesive | Pressure Sensitive |
Plate Compatibility | All Microplates |
Unit Size | Pack of 100 |
Color | Clear |
Description | Sealing Tape, -40°C - 90°C |
Material | Polyethylene, Silicone |
No. per Pack | 100 |
Product Type | Non-sterile Sealing Tape |
Product Line | Webseal |
Adhesive | Pressure Sensitive |
Temperature Range (Metric) | -40°C to +90°C |
Plate Compatibility | All Microplates |
Unit Size | Pack of 100 |
Color | Clear |
Description | Sealing Tape, -70°C - 100°C |
Material | PES, Silicone |
No. per Pack | 100 |
Product Type | Non-sterile Sealing Tape |
Product Line | Webseal |
Adhesive | Pressure Sensitive |
Temperature Range (Metric) | -70°C to +100°C |
Plate Compatibility | All Microplates |
Unit Size | Pack of 100 |
Color | Silver |
Description | Sealing Tape, -80° to 120°C |
Material | Aluminum, Silicone |
No. per Pack | 100 |
Product Type | Non-sterile Sealing Tape |
Product Line | Webseal |
Adhesive | Pressure Sensitive |
Temperature Range (Metric) | -80°C to +120°C |
Unit Size | Pack of 100 |
Color | Clear |
Description | Sealing Tape with 3 layer no adhesive at cavity area |
Material | PET, Silicone |
No. per Pack | 100 |
Product Type | Non-sterile Sealing Tape |
Product Line | Webseal |
Adhesive | Pressure Sensitive |
Plate Compatibility | All Microplates |
Unit Size | Pack of 100 |
Catalog Number | Specifications | Unit Size | Description | Material | Color | Price (USD) | Availability | Quantity | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
60180-M145 | Pack of 100 | Sealing Tape with SRA (synthetic rubber adhesive) | Polyolefin, Rubber | Clear | 366.39 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
60180-M142 | Pack of 100 | Sealing Tape, -40°C - 90°C | Polyethylene, Silicone | Clear | 503.45 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
60180-M143 | Pack of 100 | Sealing Tape, -70°C - 100°C | PES, Silicone | Clear | 253.96 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
60180-M144 | Pack of 100 | Sealing Tape, -80° to 120°C | Aluminum, Silicone | Silver | 182.67 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
60180-M146 | Pack of 100 | Sealing Tape with 3 layer no adhesive at cavity area | PET, Silicone | Clear | Special offer: 694.85 1158.08 Save 463.23 (40%) Ends: 30-Sep-2024 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|