Gas Chromatography Tubing
Product Image
Thermo Scientific™
Gas Chromatography Tubing
Find the copper and stainless steel tubing in a variety of sizes for plumbing your gas chromatography (GC) system.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberMaterialDiameter (English) InnerDiameter (English) OuterDescription
60181-632Copper0.065 in.1/8 in.50 ft. Length
60181-638Stainless Steel0.085 in.1/8 in.25 ft. Length
Catalog number 60181-632
Price (USD)
155.76
Each
Add to cart
Material:
Copper
Diameter (English) Inner:
0.065 in.
Diameter (English) Outer:
1/8 in.
Description:
50 ft. Length
Price (USD)
155.76
Each
Add to cart
Ask our AI about this Product
Find copper and stainless steel Thermo Scientific™ Gas Chromatography Tubing in a variety of sizes for plumbing your gas chromatography (GC) system.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Description50 ft. Length
Diameter (English) Inner0.065 in.
Diameter (English) Outer1/8 in.
MaterialCopper
For Use With (Equipment)Gas Chromatography Systems
TypeGas Chromatography Tubing
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link