HyperSep™ HPLC Columns for Online SPE
HyperSep™ HPLC Columns for Online SPE
Actual product may vary
Thermo Scientific™
HyperSep™ HPLC Columns for Online SPE
Clean-up and concentrate target analytes in minutes with these columns combining SPE chemistries with traditional HPLC column fittings and hardware.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSorbentDiameter (Metric) Inner
60312-401HyperSep Retain AX2.1 mm
60312-201HyperSep Retain PEP2.1 mm
60312-202HyperSep Retain PEP3.0 mm
60312-301HyperSep Retain CX2.1 mm
60312-302HyperSep Retain CX3.0 mm
60312-402HyperSep Retain AX3.0 mm
60312-501HyperSep Hypercarb2.1 mm
60312-502HyperSep Hypercarb3.0 mm
8 Options
Catalog number 60312-401
Price (USD)/ Each
536.51
Add to cart
Sorbent:
HyperSep Retain AX
Diameter (Metric) Inner:
2.1 mm
Price (USD)/ Each
536.51
Add to cart
HyperSep™ HPLC Columns for Online SPE
Catalog number60312-401
Price (USD)/ Each
536.51
-
Add to cart
Clean-up and concentrate target analytes in minutes with Thermo Scientific™ HyperSep™ Online SPE Products. Available in several convenient formats, the columns utilize popular HyperSep SPE chemistries. When used with conventional HPLC systems and appropriate switching valves, they provide:
  • Effective removal of contaminants, such as proteins, from samples
  • Fast, effective clean-up and concentration of target compounds
HyperSep HPLC Columns for Online SPE combine SPE chemistries with traditional HPLC column fittings and hardware.
HyperSep HPLC Columns for Online SPE are available with the following HyperSep SPE chemistries:
  • HyperSep Retain™ PEP — for retention of polar and nonpolar analytes
  • HyperSep Retain-CX — for retention of basic and nonpolar analytes
  • HyperSep Retain-AX — for retention of acidic and nonpolar analytes
  • HyperSep Hypercarb™ — for retention of extremely polar analytes
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
DescriptionHyperSep Retain-AX; 2.1 mm ID
For Use With (Application)Online Solid Phase Extraction
For Use With (Equipment)Conventional HPLC System
Product LineHyperSep
SorbentHyperSep Retain AX
TypeOnline HPLC Column
Diameter (Metric) Inner2.1 mm
Unit SizeEach