Vanquish™ User Interface
Thermo Scientific™
Vanquish™ User Interface
Quickly understand system status and solve problems with your HPLC system using the Thermo Scientific™ Vanquish™ user interface.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionFor Use With (Equipment)Type
6036.1170-01Vanquish User InterfaceVanquish SystemsUser Interface
6036.1180Vanquish DisplayVanquish HPLC and UHPLC SystemsTouchscreen Display Only
Catalog number 6036.1170-01
Price (USD)
-
Description:
Vanquish User Interface
For Use With (Equipment):
Vanquish Systems
Type:
User Interface
View system health, performance, and equilibration status at a glance with the Thermo Scientific™ Vanquish™ User Interface. This user-friendly interface enables operators to quickly understand system status and solve problems. The multi-language interface comes with simple maintenance tutorials to guide even novice operators in keeping the system reliably delivering results.
View HPLC/UHPLC resources ∣ HPLC Support Center
  • Glove-enabled touch display
  • Delivers insight into instrument status parameters such as flow and pressure
  • Monitors column equilibration progress through a graphical representation of pump pressure
  • Displays step-by-step maintenance videos
  • Offers remote instrument monitoring, even without installation of a CDS
  • Multi-language support
Specifications
DescriptionVanquish User Interface
For Use With (Equipment)Vanquish Systems
TypeUser Interface
Unit SizeEach