Thermo Scientific™
Viper™ Capillary Kits for Application-Specific Vanquish™ LC Systems
Catalog number: 6036.2020
Have Questions?
Thermo Scientific™
Viper™ Capillary Kits for Application-Specific Vanquish™ LC Systems
Catalog number: 6036.2020
Have Questions?
Improve chromatographic resolution by minimizing dead volume with the Thermo Scientific™ Viper™ Fingertight Fitting Systems. These pre-assembled kits allow you to quickly connect LC modules, valves, and columns for leak-free fluidic HPLC and UHPLC connections. Ideally suited for running application-specific workflows with your HPLC and LC-MS systems, like the Thermo Scientific™ Vanquish™ Method Development LC Systems, Thermo Scientific Vanquish Online 2D-LC Systems, or Thermo Scientific Vanquish Duo UHPLC Systems.
See Viper and nanoViper resources
Catalog Number
6036.2020
Unit Size
Each
Description
Viper Kit for Vanquish Duo for Tandem LC and LC-MS
For Use With (Application)
Intelligent column switching for maximized detector usage
For Use With (Equipment)
Vanquish Flex UHPLC Systems, Vanquish Horizon UHPLC Systems
Type
Advanced Capillary Kit
Price (USD)
Availability
-
Full specifications
Quantity
DescriptionViper Kit for Vanquish Duo for Tandem LC and LC-MS
For Use With (Application)Intelligent column switching for maximized detector usage
For Use With (Equipment)Vanquish Flex UHPLC Systems, Vanquish Horizon UHPLC Systems
MaterialMP35N
Max. Pressure1500 bar
Product LineViper
TypeAdvanced Capillary Kit
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Extension Kit for Automated Method Scouting for Vanquish LC Systems
For Use With (Application)
Method Development
For Use With (Equipment)
Vanquish Core HPLC System, Vanquish Flex UHPLC Systems, Vanquish Horizon UHPLC Systems
Type
Extension Kit
Price (USD)
Availability
-
Full specifications
Quantity
DescriptionExtension Kit for Automated Method Scouting for Vanquish LC Systems
For Use With (Application)Method Development
For Use With (Equipment)Vanquish Core HPLC System, Vanquish Flex UHPLC Systems, Vanquish Horizon UHPLC Systems
TypeExtension Kit
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Viper Kit for Trap Heart-Cut and Simple Switch 2D-LC for Vanquish UHPLC Systems
For Use With (Application)
2D-LC
For Use With (Equipment)
Vanquish Flex UHPLC Systems, Vanquish Horizon UHPLC Systems
Type
Advanced Capillary Kit
Price (USD)
Availability
-
Full specifications
Quantity
DescriptionViper Kit for Trap Heart-Cut and Simple Switch 2D-LC for Vanquish UHPLC Systems
For Use With (Application)2D-LC
For Use With (Equipment)Vanquish Flex UHPLC Systems, Vanquish Horizon UHPLC Systems
MaterialMP35N
Max. Pressure1500 bar
Product LineViper
TypeAdvanced Capillary Kit
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Viper Kit for Vanquish Duo for Dual LC
For Use With (Application)
Two of the same of two different assays simultaneously
For Use With (Equipment)
Vanquish Flex UHPLC Systems, Vanquish Horizon UHPLC Systems
Type
Advanced Capillary Kit
Price (USD)
Availability
-
Full specifications
Quantity
DescriptionViper Kit for Vanquish Duo for Dual LC
For Use With (Application)Two of the same of two different assays simultaneously
For Use With (Equipment)Vanquish Flex UHPLC Systems, Vanquish Horizon UHPLC Systems
MaterialMP35N
Max. Pressure1500 bar
Product LineViper
TypeAdvanced Capillary Kit
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Viper Kit for Vanquish Duo for Inverse Gradient
For Use With (Application)
Standard- free quantitation with charged aerosol detection
For Use With (Equipment)
Vanquish Flex UHPLC Systems, Vanquish Horizon UHPLC Systems
Type
Advanced Capillary Kit
Price (USD)
Availability
-
Full specifications
Quantity
DescriptionViper Kit for Vanquish Duo for Inverse Gradient
For Use With (Application)Standard- free quantitation with charged aerosol detection
For Use With (Equipment)Vanquish Flex UHPLC Systems, Vanquish Horizon UHPLC Systems
MaterialMP35N
Max. Pressure1500 bar
Product LineViper
TypeAdvanced Capillary Kit
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Viper Kit for Automated Method Scouting for Vanquish UHPLC Systems
For Use With (Application)
Method Development
For Use With (Equipment)
Vanquish Flex and Horizon UHPLC Systems
Type
Advanced Capillary Kit
Price (USD)
Availability
-
Full specifications
Quantity
DescriptionViper Kit for Automated Method Scouting for Vanquish UHPLC Systems
For Use With (Application)Method Development
For Use With (Equipment)Vanquish Flex and Horizon UHPLC Systems
MaterialMP35N
Max. Pressure1500 bar
Product LineViper
TypeAdvanced Capillary Kit
Length (Metric)Various
Diameter (Metric) Outer100 μm
Unit SizeEach
Showing 6 of 6
Catalog NumberSpecificationsUnit SizeDescriptionFor Use With (Application)For Use With (Equipment)TypePrice (USD)AvailabilityQuantity
6036.2020Full specifications
EachViper Kit for Vanquish Duo for Tandem LC and LC-MSIntelligent column switching for maximized detector usageVanquish Flex UHPLC Systems, Vanquish Horizon UHPLC SystemsAdvanced Capillary Kit
-
DescriptionViper Kit for Vanquish Duo for Tandem LC and LC-MS
For Use With (Application)Intelligent column switching for maximized detector usage
For Use With (Equipment)Vanquish Flex UHPLC Systems, Vanquish Horizon UHPLC Systems
MaterialMP35N
Max. Pressure1500 bar
Product LineViper
TypeAdvanced Capillary Kit
Unit SizeEach
6036.0100Full specifications
EachExtension Kit for Automated Method Scouting for Vanquish LC SystemsMethod DevelopmentVanquish Core HPLC System, Vanquish Flex UHPLC Systems, Vanquish Horizon UHPLC SystemsExtension Kit
-
DescriptionExtension Kit for Automated Method Scouting for Vanquish LC Systems
For Use With (Application)Method Development
For Use With (Equipment)Vanquish Core HPLC System, Vanquish Flex UHPLC Systems, Vanquish Horizon UHPLC Systems
TypeExtension Kit
Unit SizeEach
6036.2030Full specifications
EachViper Kit for Trap Heart-Cut and Simple Switch 2D-LC for Vanquish UHPLC Systems2D-LCVanquish Flex UHPLC Systems, Vanquish Horizon UHPLC SystemsAdvanced Capillary Kit
-
DescriptionViper Kit for Trap Heart-Cut and Simple Switch 2D-LC for Vanquish UHPLC Systems
For Use With (Application)2D-LC
For Use With (Equipment)Vanquish Flex UHPLC Systems, Vanquish Horizon UHPLC Systems
MaterialMP35N
Max. Pressure1500 bar
Product LineViper
TypeAdvanced Capillary Kit
Unit SizeEach
6036.2305Full specifications
EachViper Kit for Vanquish Duo for Dual LCTwo of the same of two different assays simultaneouslyVanquish Flex UHPLC Systems, Vanquish Horizon UHPLC SystemsAdvanced Capillary Kit
-
DescriptionViper Kit for Vanquish Duo for Dual LC
For Use With (Application)Two of the same of two different assays simultaneously
For Use With (Equipment)Vanquish Flex UHPLC Systems, Vanquish Horizon UHPLC Systems
MaterialMP35N
Max. Pressure1500 bar
Product LineViper
TypeAdvanced Capillary Kit
Unit SizeEach
6036.2010Full specifications
EachViper Kit for Vanquish Duo for Inverse GradientStandard- free quantitation with charged aerosol detectionVanquish Flex UHPLC Systems, Vanquish Horizon UHPLC SystemsAdvanced Capillary Kit
-
DescriptionViper Kit for Vanquish Duo for Inverse Gradient
For Use With (Application)Standard- free quantitation with charged aerosol detection
For Use With (Equipment)Vanquish Flex UHPLC Systems, Vanquish Horizon UHPLC Systems
MaterialMP35N
Max. Pressure1500 bar
Product LineViper
TypeAdvanced Capillary Kit
Unit SizeEach
6036.2807Full specifications
EachViper Kit for Automated Method Scouting for Vanquish UHPLC SystemsMethod DevelopmentVanquish Flex and Horizon UHPLC SystemsAdvanced Capillary Kit
-
DescriptionViper Kit for Automated Method Scouting for Vanquish UHPLC Systems
For Use With (Application)Method Development
For Use With (Equipment)Vanquish Flex and Horizon UHPLC Systems
MaterialMP35N
Max. Pressure1500 bar
Product LineViper
TypeAdvanced Capillary Kit
Length (Metric)Various
Diameter (Metric) Outer100 μm
Unit SizeEach
Showing 6 of 6
Enjoy robust and reliable fingertight fitting LC connections. Tailored to your advanced workflow requirements, Viper application-specific capillary kits offer:
  • Virtually zero-dead volume fingertight connections
  • Easy-to-install, tool-free reusable capillaries
  • Reliable, reproducible LC connections that deliver low carryover and simplify method transfer
  • Support for a wide range of applications with different materials and operating pressures up to 1517 bar (22,000 psi)