Column Switching Valve for UltiMate™ 3000 RSLC Nano Systems
Column Switching Valve for UltiMate™ 3000 RSLC Nano Systems
Actual product may vary
Thermo Scientific™
Column Switching Valve for UltiMate™ 3000 RSLC Nano Systems
Column Switching Valve for UltiMate 3000 RSLC Nano Systems offer convenience and confidence for column installation.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberStyle
6041.00042-Position; 6-Port
6041.00012-Position; 10-Port
6041.00122-Position; 10-Port; PEEK
3 Options
Catalog number 6041.0004
Price (USD)/ Each
-
Request A Quote
Style:
2-Position; 6-Port
Offer convenience and confidence for column installation with Thermo Scientific™ Dionex Column Switching Valve for UltiMate™ 3000 RSLC Nano Systems.
  • Low-dispersion, two-port valves
  • Port-to-port volume 124nL
Specifications
DescriptionColumn switching valve; 2-position, 6-port valve
For Use With (Equipment)RSLC Nano NCS-3500
MaterialStainless Steel
Max. Pressure13050 psi (900 bar)
Product LineUltiMate
TypeNano/Cap Column Switching Valve
Style2-Position; 6-Port
Unit SizeEach