Invitrogen™

Platinum™ Tfi Exo(-) DNA Polymerase

Catalog number: 60684050
Have Questions?
Invitrogen™

Platinum™ Tfi Exo(-) DNA Polymerase

Catalog number: 60684050
Have Questions?
Catalog Number
60684050
Unit Size
500 units
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
60684050500 units
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Platinum™ Tfi Exo(-) DNA Polymerase is recombinant Tfi Exo(-) DNA Polymerase complexed with a proprietary antibody mix that inhibits polymerase activity at ambient temperature, allowing room-temperature reaction setup. Activity is restored after the initial denaturation step in PCR cycling at 94°C, providing an automatic 'hot start' for increased specificity, sensitivity, and yield. Tfi Exo(-) DNA Polymerase is purified from E. coli. expressing cloned mutants of the Thermus filiformis DNA polymerase gene. This enzyme has 5'→3' polymerase activity, but lacks both 5'→3' and 3'→5' exonuclease activity for improved yield and robustness. Tfi Exo(-) DNA polymerase is heat-stable and will synthesize DNA at elevated temperatures from single-stranded templates in the presence of a primer.

Applications
Platinum™ Tfi Exo(-) DNA Polymerase can be used in protocols that currently use Platinum™ Taq DNA Polymerase without modification. PCR performance is comparable to that of Platinum™ Taq in yield, specificity, fidelity, and robustness. Like Platinum™ Taq, Platinum™ Tfi Exo(-) DNA Polymerase has a nontemplate-dependent terminal transferase activity that adds a single deoxyadenosine (A) to the 3' ends of PCR products.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

GC-Rich PCR Performance
Low
Polymerase
Tfi DNA Polymerase
Reaction Speed
Standard
Product Line
Platinum™
Product Type
DNA Polymerase
Shipping Condition
Dry Ice
Concentration
5X
Fidelity (vs. Taq)
1 X
Hot Start
Built-In Hot Start
Overhang
3'-A
Reaction Format
Separate Components
Size (Final Product)
1 kb or less
Volume
5.2 mL

Contents & Storage

Platinum™ Tfi DNA Polymerase (500 units total, at 5U/μL) is supplied with 5X Platinum™ Tfi Reaction Buffer (4 × 1.3 mL) and 50 mM Magnesium Chloride (1 mL). Store at -20°C (non-frost-free).

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text