Search Thermo Fisher Scientific
Description | PCM-3000 pH and Conductivity Monitor for DAD-3000/MWD-3000 Series |
Detection Mode | UV-vis Detection |
Type | pH and Conductivity Monitor |
For Use With (Equipment) | DAD-3000 and MWD-3000 |
Unit Size | Each |
Description | PCM-3000 pH and Conductivity Monitor for VWD-3 x 00 Series |
Detection Mode | UV-vis Detection |
Type | pH and Conductivity Monitor |
For Use With (Equipment) | VWD-3x00 |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | For Use With (Equipment) | Price (USD) | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
6082.2000 | Each | DAD-3000 and MWD-3000 | Contact Us › | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
6082.2005 | Each | VWD-3x00 | Contact Us › | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
The PCM-3000 can be used for monitoring gradient formation in salt- or pH-gradient-based ion-exchange chromatography (IEC), typically used for biopharmaceutical analysis.
pH gradients are often used for proteins and large molecules where polarity differences alone can’t resolve different species with a typical organic gradient.
IEC is a versatile separation technique for profiling the charge heterogeneity of biotherapeutic proteins, including monoclonal antibodies (MAbs). The elution can be driven by pH, ionic strength, or a combination of these two parameters. The PCM-3000 can also be used to find the optimal elution conditions for specific proteins and MAbs.
Features