1-Butanol, HPLC Grade 99.4%, Thermo Scientific Chemicals
1-Butanol, HPLC Grade 99.4%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Butanol, HPLC Grade 99.4%, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
4 L
Packaging:
EcoSafPak™
Catalog number 610020040
Price (USD)/ Each
304.65
Online exclusive
338.00 
Save 33.35 (10%)
-
Add to cart
Quantity:
4 L
Packaging:
EcoSafPak™
Request bulk or custom format
Price (USD)/ Each
304.65
Online exclusive
338.00 
Save 33.35 (10%)
Add to cart
1-Butanol, HPLC Grade 99.4%, Thermo Scientific Chemicals
Catalog number610020040
Price (USD)/ Each
304.65
Online exclusive
338.00 
Save 33.35 (10%)
-
Add to cart
Chemical Identifiers
IUPAC Namebutan-1-ol
Molecular FormulaC4H10O
InChI KeyLRHPLDYGYMQRHN-UHFFFAOYSA-N
SMILESCCCCO
Molecular Weight (g/mol)74.12
View more
SpecificationsSpecification SheetSpecification Sheet
UVat 254 nm A: =<0.025
Color scale=<10 APHA
Identificationpasses test
Refractive index1.3952 to 1.3992 (25°C, 589 nm)
GC>=99.4 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for HPLC
RUO – Research Use Only