Thermo Scientific Chemicals

Hexanes, for HPLC, Thermo Scientific Chemicals

Catalog number: 610070040
4 L, EcoSafPak™, Each
Thermo Scientific Chemicals

Hexanes, for HPLC, Thermo Scientific Chemicals

Catalog number: 610070040
4 L, EcoSafPak™, Each
Quantity
Packaging
Catalog number: 610070040
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
92112-69-1, 110-54-3
UV
1 cm cell vs water
UV
at 195 nm A: =<1
UV
at 210 nm A: =<0.25
UV
at 254 nm A: =<0.014
Specific gravity
0.653 to 0.673

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For HPLC
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text