1x1 image pixel for data collection
Methanol, 99.9%, for HPLC
Methanol, 99.9%, for HPLC
Methanol, 99.9%, for HPLC
Methanol, 99.9%, for HPLC
Thermo Scientific Chemicals

Methanol, 99.9%, for HPLC

CAS: 67-56-1 | CH4O | 32.04 g/mol
Have Questions?
Catalog NumberQuantityPackaging
6100900404 LEcoSafPak™
Catalog number 610090040
Price (USD)
182.00
Each
In stock
Add to cart
Quantity:
4 L
Packaging:
EcoSafPak™
Request bulk or custom format
Price (USD)
182.00
Each
Add to cart
Chemical Identifiers
CAS67-56-1
IUPAC Namemethanol
Molecular FormulaCH4O
InChI KeyOKKJLVBELUTLKV-UHFFFAOYSA-N
SMILESCO
View more
SpecificationsSpecification SheetSpecification Sheet
Titrable base=<0.0002 meq/g
Residue after evaporation=<3 ppm
UV1 cm cell vs water
UVat 205 nm A: =<1
UVat 220 nm A: =<0.3
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For HPLC
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection