Dichloromethane, for residue analysis, Thermo Scientific Chemicals
Dichloromethane, for residue analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Dichloromethane, for residue analysis, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
4 L
Packaging:
EcoSafPak™
Catalog number 610160040
Price (USD)/ Each
214.65
Online exclusive
238.00 
Save 23.35 (10%)
-
Add to cart
Quantity:
4 L
Packaging:
EcoSafPak™
Request bulk or custom format
Price (USD)/ Each
214.65
Online exclusive
238.00 
Save 23.35 (10%)
Add to cart
Dichloromethane, for residue analysis, Thermo Scientific Chemicals
Catalog number610160040
Price (USD)/ Each
214.65
Online exclusive
238.00 
Save 23.35 (10%)
-
Add to cart
Chemical Identifiers
CAS75-09-2
IUPAC Namedichloromethane
Molecular FormulaCH2Cl2
InChI KeyYMWUJEATGCHHMB-UHFFFAOYSA-N
SMILESClCCl
View more
SpecificationsSpecification SheetSpecification Sheet
Additional infopesticide residue analysis < 20 ng/l
Appearance (Form)Clear liquid
Color scale=<10 APHA
Infrared spectrumConforms
Amylene40 to 100 ppm
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For residue analysis
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only