tert-Butyl MethyleEther, 99%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
tert-Butyl MethyleEther, 99%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals

tert-Butyl MethyleEther, 99%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
100 mL
Catalog number 610271000
Price (USD)/ Each
58.65
Online exclusive
64.50 
Save 5.85 (9%)
-
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (USD)/ Each
58.65
Online exclusive
64.50 
Save 5.85 (9%)
Add to cart
tert-Butyl MethyleEther, 99%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
Catalog number610271000
Price (USD)/ Each
58.65
Online exclusive
64.50 
Save 5.85 (9%)
-
Add to cart
Chemical Identifiers
CAS1634-04-4
IUPAC Name2-methoxy-2-methylpropane
Molecular FormulaC5H12O
InChI KeyBZLVMXJERCGZMT-UHFFFAOYSA-N
SMILESCOC(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless
Appearance (Form)Liquid
Assay>=99 %
Water=<50 ppm
Peroxides=<1 ppm (as H2O2)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only