Chloroform, 99.9%, Extra Dry, stabilized, AcroSeal™, Thermo Scientific Chemicals
Chloroform, 99.9%, Extra Dry, stabilized, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Chloroform, 99.9%, Extra Dry, stabilized, AcroSeal™, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle in Tin Can
Catalog number 610281000
Price (USD)/ Each
46.65
Online exclusive
51.90 
Save 5.25 (10%)
-
Add to cart
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle in Tin Can
Request bulk or custom format
Price (USD)/ Each
46.65
Online exclusive
51.90 
Save 5.25 (10%)
Add to cart
Chloroform, 99.9%, Extra Dry, stabilized, AcroSeal™, Thermo Scientific Chemicals
Catalog number610281000
Price (USD)/ Each
46.65
Online exclusive
51.90 
Save 5.25 (10%)
-
Add to cart
Chemical Identifiers
CAS67-66-3
IUPAC Nametrichloromethane
Molecular FormulaCHCl3
InChI KeyHEDRZPFGACZZDS-UHFFFAOYSA-N
SMILESClC(Cl)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Water=<50 ppm
Appearance (Color)Colorless
Appearance (Form)Liquid
Assay>=99.9 %
Additional infoinhibitor: 40 - 100 ppm
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only