Thermo Scientific Chemicals

N,N-Dimethylformamide, 99.8%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 610321000
100 mL, Each
Thermo Scientific Chemicals

N,N-Dimethylformamide, 99.8%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 610321000
100 mL, Each
Quantity
Catalog number: 610321000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
68-12-2
IUPAC Name
N,N-dimethylformamide
Molecular Formula
C3H7NO
InChI Key
ZMXDDKWLCZADIW-UHFFFAOYSA-N
SMILES
CN(C)C=O
Appearance (Color)
Colorless
Appearance (Form)
Liquid
Assay
>=99.8 %
Water
=<50 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text