Thermo Scientific Chemicals

Ethyl acetate, 99.9%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 610341000
100 mL, Each
Thermo Scientific Chemicals

Ethyl acetate, 99.9%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 610341000
100 mL, Each
Quantity
Catalog number: 610341000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
141-78-6
IUPAC Name
ethyl acetate
Molecular Formula
C4H8O2
InChI Key
XEKOWRVHYACXOJ-UHFFFAOYSA-N
SMILES
CCOC(C)=O
Appearance (Color)
Colorless
Appearance (Form)
Liquid
Assay
>=99.9 %
Water
=<50 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text