Thermo Scientific Chemicals

Methyl sulfoxide, 99.7%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 610421000
100 mL, Each
Thermo Scientific Chemicals

Methyl sulfoxide, 99.7%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 610421000
100 mL, Each
Quantity
Catalog number: 610421000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
67-68-5
IUPAC Name
methanesulfinylmethane
Molecular Formula
C2H6OS
InChI Key
IAZDPXIOMUYVGZ-UHFFFAOYSA-N
SMILES
CS(C)=O
Assay
>=99.7 %
Water
=<50 ppm
Appearance (Color)
Colorless
Appearance (Form)
Liquid
Identification
passes test

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text