Isopropanol, 99.5%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
Isopropanol, 99.5%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Isopropanol, 99.5%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle in Tin Can
Catalog number 610431000
Price (USD)/ Each
46.80
-
Add to cart
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle in Tin Can
Request bulk or custom format
Price (USD)/ Each
46.80
Add to cart
Isopropanol, 99.5%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
Catalog number610431000
Price (USD)/ Each
46.80
-
Add to cart
Chemical Identifiers
CAS67-63-0
IUPAC Namepropan-2-ol
Molecular FormulaC3H8O
InChI KeyKFZMGEQAYNKOFK-UHFFFAOYSA-N
SMILESCC(C)O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless
Assay>=99.5 %
Water=<50 ppm
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only