Dichloromethane, 99.9%, Extra Dry, anhydrous, SC, AcroSeal™, Thermo Scientific Chemicals
Dichloromethane, 99.9%, Extra Dry, anhydrous, SC, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Dichloromethane, 99.9%, Extra Dry, anhydrous, SC, AcroSeal™, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle in Tin Can
Catalog number 610931000
Price (USD)/ Each
53.65
Online exclusive
59.00 
Save 5.35 (9%)
-
Add to cart
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle in Tin Can
Request bulk or custom format
Price (USD)/ Each
53.65
Online exclusive
59.00 
Save 5.35 (9%)
Add to cart
Dichloromethane, 99.9%, Extra Dry, anhydrous, SC, AcroSeal™, Thermo Scientific Chemicals
Catalog number610931000
Price (USD)/ Each
53.65
Online exclusive
59.00 
Save 5.35 (9%)
-
Add to cart
Chemical Identifiers
CAS75-09-2
IUPAC Namedichloromethane
Molecular FormulaCH2Cl2
InChI KeyYMWUJEATGCHHMB-UHFFFAOYSA-N
SMILESClCCl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless
Appearance (Form)Liquid
Assay>=99.9 %
Water=<50 ppm (K.F.)
Additional infoinhibitor: 40 - 100 ppm amylene
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only