Acetonitrile, 99.9%, Extra Dry, anhydrous, SC, AcroSeal™, Thermo Scientific Chemicals
Acetonitrile, 99.9%, Extra Dry, anhydrous, SC, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Acetonitrile, 99.9%, Extra Dry, anhydrous, SC, AcroSeal™, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
100 mL
Packaging:
AcroSeal™
Catalog number 610961000
Price (USD)/ Each
69.65
Online exclusive
82.00 
Save 12.35 (15%)
-
Add to cart
Quantity:
100 mL
Packaging:
AcroSeal™
Request bulk or custom format
Price (USD)/ Each
69.65
Online exclusive
82.00 
Save 12.35 (15%)
Add to cart
Acetonitrile, 99.9%, Extra Dry, anhydrous, SC, AcroSeal™, Thermo Scientific Chemicals
Catalog number610961000
Price (USD)/ Each
69.65
Online exclusive
82.00 
Save 12.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Nameacetonitrile
Molecular FormulaC2H3N
InChI KeyWEVYAHXRMPXWCK-UHFFFAOYSA-N
SMILESCC#N
Molecular Weight (g/mol)41.05
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless
Appearance (Form)Liquid
Identificationpasses test
Assay>=99.9 %
Water=<50 ppm (K.F.)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only