1x1 image pixel for data collection
Pyridine, 99.0%, Extra Dry, anhydrous, SC, AcroSeal™, Thermo Scientific Chemicals
Pyridine, 99.0%, Extra Dry, anhydrous, SC, AcroSeal™, Thermo Scientific Chemicals
Pyridine, 99.0%, Extra Dry, anhydrous, SC, AcroSeal™, Thermo Scientific Chemicals
Pyridine, 99.0%, Extra Dry, anhydrous, SC, AcroSeal™, Thermo Scientific Chemicals
Pyridine, 99.0%, Extra Dry, anhydrous, SC, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Pyridine, 99.0%, Extra Dry, anhydrous, SC, AcroSeal™, Thermo Scientific Chemicals

 
Have Questions?
Catalog NumberQuantity
610991000100 mL
Catalog number 610991000
Price (USD)
-
Quantity:
100 mL
Request bulk or custom format
Chemical Identifiers
CAS110-86-1
IUPAC Namepyridine
Molecular FormulaC5H5N
InChI KeyJUJWROOIHBZHMG-UHFFFAOYSA-N
SMILESC1=CC=NC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=99.0 %
Water=<50 ppm
Appearance (Form)Liquid
Identificationpasses test
Appearance (Color)Colorless
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link