Informational:Informational:Save up to 31% on your order of cell culture products. View promotion details »
Gibco™

MEM, powder

Catalog number: 61100087
Have Questions?
Gibco™

MEM, powder

Catalog number: 61100087
Have Questions?
Catalog Number
61100087
Unit Size
10 L
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
10 x 1 L
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
50 L
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
6110008710 L
-
6110006110 x 1 L
-
6110010350 L
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations

Minimum Essential Medium (MEM) is one of the most commonly used of all cell culture media. MEM can be used with a variety of suspension and adherent mammalian cells, including HeLa, BHK-21, 293, HEP-2, HT-1080, MCF-7, fibroblasts, and primary rat astrocytes. We offer a variety of Gibco™ MEM modifications for a range of cell culture applications. Find the right formulation using the media selector tool.


This MEM is modified as follows:
WithWithout
• L-glutamine• HEPES
• Phenol Red• Sodium Bicarbonate


The complete formulation is available.

Gibco™ MEM, developed by Harry Eagle, was based on his earlier formulation of Basal Medium Eagle (BME). Many other modifications of MEM followed, including Glasgow’s MEM, MEM α, DMEM, and Temin’s Modification. MEM is available with Earle’s salts for use in a CO2 incubator, or with Hanks' salts for use without CO2. This product is made with Earle’s salts.

Using MEM
MEM contains no proteins, lipids, or growth factors. Therefore, MEM requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). MEM uses a sodium bicarbonate buffer system (2.2 g/L) and therefore requires a 5–10% CO2 environment to maintain physiological pH. Powder forms of Gibco™ cell culture medium require sodium bicarbonate supplementation, pH adjustment, and filtration at the time of preparation (see protocol for details).

Specifications

Cell Line
HeLa, BHK-21, 293, HEP-2, HT-1080, MCF-7, and fibroblasts
Cell Type
Primary Rat Astrocytes
Classification
Animal Origin-free
Form
Powder
Product Type
MEM (Minimum Essential Medium)
Serum Level
Standard Serum Supplementation
Shelf Life
36 Months From Date of Manufacture
Sterility
Sterile-filtered
With Additives
Low Glucose, Glutamine, Phenol Red
Without Additives
No HEPES, No Sodium Pyruvate, No Sodium Bicarbonate
Manufacturing Quality
cGMP-compliant under the ISO 13485 standard
Product Line
Gibco™
Quantity
10 L
Shipping Condition
Room Temperature

Contents & Storage

Storage conditions: 2-8° C
Shipping conditions: Ambient
Shelf life: 36 months from date of manufacture

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text