Thermo Scientific Chemicals

Buffer Solution, pH 10, Color-coded blue, Certified, Thermo Scientific Chemicals

Catalog number: 611020040
4 L, Each
Thermo Scientific Chemicals

Buffer Solution, pH 10, Color-coded blue, Certified, Thermo Scientific Chemicals

Catalog number: 611020040
4 L, Each
Quantity
Catalog number: 611020040
Price (USD)
Quantity
-
Have Questions?
Appearance (Color)
Clear blue
Appearance (Form)
Liquid
Identification
passes test
pH
9.98 to 10.02 (at 25°C)
Additional info
optcal absorbance: passes test

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text