1x1 image pixel for data collection
Buffer solution, pH 7.00 , Color-coded yellow, Certified
Buffer solution, pH 7.00 , Color-coded yellow, Certified
Buffer solution, pH 7.00 , Color-coded yellow, Certified
Buffer solution, pH 7.00 , Color-coded yellow, Certified
Thermo Scientific Chemicals

Buffer solution, pH 7.00 , Color-coded yellow, Certified

CAS: 7778-77-0 | H2KO4P | 136.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
4 L
Catalog number 611065000
Price (USD)
61.30
Each
In stock
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (USD)
61.30
Each
Add to cart
Specifications
AbsorbancePass Test
CAS7778-77-0
ChEBICHEBI:63036
Chemical Name or MaterialBuffer solution pH 7.00
IUPAC Namepotassium dihydrogen phosphate
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection