Thermo Scientific Chemicals

Malachite Green

Catalog number: 611255000
500 g, Each
Thermo Scientific Chemicals

Malachite Green

Catalog number: 611255000
500 g, Each
Quantity
Catalog number: 611255000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
569-64-2
IUPAC Name
bis[4-(dimethylamino)phenyl](phenyl)methylium chloride
Molecular Formula
C23H25ClN2
InChI Key
FDZZZRQASAIRJF-UHFFFAOYSA-M
SMILES
[Cl-].CN(C)C1=CC=C(C=C1)[C+](C1=CC=CC=C1)C1=CC=C(C=C1)N(C)C
Identification
Passes test
pH
Passes test
Appearance (Color)
Green
Appearance (Form)
Crystals

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text