Thermo Scientific Chemicals

Dextran Sulfate Sodium Salt

Catalog number: 611361000
100 g, Each
Thermo Scientific Chemicals

Dextran Sulfate Sodium Salt

Catalog number: 611361000
100 g, Each
Quantity
Catalog number: 611361000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
9011-18-1
IUPAC Name
Dextran sulfate sodium salt
MDL Number
MFCD00081551
Appearance (Color)
White to off-white
Appearance (Form)
Powder
Identification
passes test
Loss on drying
=<7.5 % ()
Additional info
sulfur: 17 - 20 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text