Nickel(II) Carbonate, Thermo Scientific Chemicals
Nickel(II) Carbonate, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Nickel(II) Carbonate, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number 611481000
Price (USD)/ Each
-
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS3333-67-3
IUPAC Namenickel(2+) carbonate
Molecular FormulaCNiO3
InChI KeyZULUUIKRFGGGTL-UHFFFAOYSA-L
SMILES[Ni++].[O-]C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Light green
Identificationpasses test
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only