Thermo Scientific Chemicals

Zinc stearate, Thermo Scientific Chemicals

Catalog number: 611515000
500 g, Each
Thermo Scientific Chemicals

Zinc stearate, Thermo Scientific Chemicals

Catalog number: 611515000
500 g, Each
Quantity
Catalog number: 611515000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
557-05-1
IUPAC Name
zinc(2+) dioctadecanoate
Molecular Formula
C36H70O4Zn
InChI Key
XOOUIPVCVHRTMJ-UHFFFAOYSA-L
SMILES
[Zn++].CCCCCCCCCCCCCCCCCC([O-])=O.CCCCCCCCCCCCCCCCCC([O-])=O
Appearance (Color)
White
Organic volatile imp.
meets requirements
Additional info
alkalies and alkaline earts: < 1.0%
Appearance (Form)
Powder
Identification
Passes test

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text