Tritolyl Phosphate, Thermo Scientific Chemicals
Tritolyl Phosphate, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tritolyl Phosphate, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
1 L
Packaging:
EcoSafPak™
Catalog number 611550010
Price (USD)/ Each
125.45
-
Add to cart
Quantity:
1 L
Packaging:
EcoSafPak™
Request bulk or custom format
Price (USD)/ Each
125.45
Add to cart
Tritolyl Phosphate, Thermo Scientific Chemicals
Catalog number611550010
Price (USD)/ Each
125.45
-
Add to cart
Chemical Identifiers
CAS1330-78-5
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear viscous liquid
Identificationpasses test
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only