Sodium Sulfate Decahydrate, Thermo Scientific Chemicals
Sodium Sulfate Decahydrate, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sodium Sulfate Decahydrate, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
500 g
Catalog number 611615000
Price (USD)/ Each
-
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
IUPAC Namedisodium decahydrate sulfate
Molecular FormulaH20Na2O14S
InChI KeyRSIJVJUOQBWMIM-UHFFFAOYSA-L
SMILESO.O.O.O.O.O.O.O.O.O.[Na+].[Na+].[O-]S([O-])(=O)=O
Molecular Weight (g/mol)322.19
View more
SpecificationsSpecification SheetSpecification Sheet
IdentificationPasses test
Free alkalinone
Heavy metals (as Pb)=<5 ppm
Chloride (Cl)=<0.001 %
Phosphate (PO4)=<0.0015 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only