Thermo Scientific Chemicals

Fuller's Earth, Thermo Scientific Chemicals

Catalog number: 611710030
3 kg, Each
Thermo Scientific Chemicals

Fuller's Earth, Thermo Scientific Chemicals

Catalog number: 611710030
3 kg, Each
Quantity
Catalog number: 611710030
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
8031-18-3
Appearance (Color)
Light yellow to brown
Appearance (Form)
Powder
Identification
passes test

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text