Thermo Scientific Chemicals

Potassium Citrate Monohydrate, Thermo Scientific Chemicals

Catalog number: 611755000
500 g, Each
Thermo Scientific Chemicals

Potassium Citrate Monohydrate, Thermo Scientific Chemicals

Catalog number: 611755000
500 g, Each
Quantity
Catalog number: 611755000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6100-05-6
IUPAC Name
tripotassium 2-hydroxypropane-1,2,3-tricarboxylate hydrate
Molecular Formula
C6H7K3O8
InChI Key
PJAHUDTUZRZBKM-UHFFFAOYSA-K
SMILES
O.[K+].[K+].[K+].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O
Identification
Passes test
Insoluble matter
=<0.01 %
Calcium (Ca)
=<0.005 %
Iron (Fe)
=<0.001 %
Sulfate (SO4)
=<0.010 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text