Aphidicolin
Thermo Scientific Chemicals

Aphidicolin

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mg
1 mg
Catalog number 611970050
Price (USD)/ Each
446.00
-
Add to cart
Quantity:
5 mg
Request bulk or custom format
Price (USD)/ Each
446.00
Add to cart
Aphidicolin
Catalog number611970050
Price (USD)/ Each
446.00
-
Add to cart
Chemical Identifiers
CAS38966-21-1
IUPAC Name(1S,2S,5R,6R,7R,10S,12R,13R)-6,13-bis(hydroxymethyl)-2,6-dimethyltetracyclo[10.3.1.0¹,¹⁰.0²,⁷]hexadecane-5,13-diol
Molecular FormulaC20H34O4
InChI KeyNOFOAYPPHIUXJR-APNQCZIXSA-N
SMILESC[C@@]1(CO)[C@H](O)CC[C@@]2(C)[C@H]1CC[C@H]1C[C@@H]3C[C@]21CC[C@]3(O)CO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Elemental Analysis9.92 to 10.32 % (H)
IdentificationPasses test
TLC>=98 %
Melting point226°C to 235°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only