Thermo Scientific Chemicals

Tris Buffer, 2M Solution

Catalog number: 612021000
100 mL, Each
Thermo Scientific Chemicals

Tris Buffer, 2M Solution

Catalog number: 612021000
100 mL, Each
Quantity
Catalog number: 612021000
Price (USD)
Quantity
-
Have Questions?
Optical density
=<0.05 (280 nm, 1 M, H2O)
pH
10.1 to 11.1 (25°C)
Copper (Cu)
=<1 ppm
Magnesium (Mg)
=<5 ppm
Additional info
DNASE: not detected

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text