Thermo Scientific Chemicals

Glycogen

Catalog number: 612030050
5 g, Each
Thermo Scientific Chemicals

Glycogen

Catalog number: 612030050
5 g, Each
Quantity
Catalog number: 612030050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
9005-79-2
IUPAC Name
(2S,3R,4S,5S,6R)-2-{[(2R,3S,4R,5R,6R)-4,5-dihydroxy-6-{[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}-3-{[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}oxan-2-yl]methoxy}-6-(hydroxymethyl)oxane-3,4,5-triol
Molecular Formula
C24H42O21
InChI Key
BYSGBSNPRWKUQH-UJDJLXLFSA-N
SMILES
OC[C@H]1O[C@H](OC[C@H]2O[C@H](O[C@H]3[C@H](O)[C@@H](O)[C@@H](O)O[C@@H]3CO)[C@H](O)[C@@H](O)[C@@H]2O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@H](O)[C@@H](O)[C@@H]1O
Specific optical rotation
165° to 185° (20°C, 589 nm)
Identification
passes test
TLC
one spot
Appearance (Color)
White to straw yellow
Appearance (Form)
Crystals

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text