Chemical Identifiers
CAS10124-56-8
IUPAC Namehexasodium hexaoxocyclohexaphosphoxane-2,4,6,8,10,12-hexakis(olate)
Molecular FormulaNa6O18P6
InChI KeyGCLGEJMYGQKIIW-UHFFFAOYSA-H
SMILES[Na+].[Na+].[Na+].[Na+].[Na+].[Na+].[O-]P1(=O)OP([O-])(=O)OP([O-])(=O)OP([O-])(=O)OP([O-])(=O)OP([O-])(=O)O1
View more