Barium Chloride, Anhydrous, Thermo Scientific Chemicals
Barium Chloride, Anhydrous, Thermo Scientific Chemicals
Barium Chloride, Anhydrous, Thermo Scientific Chemicals
Barium Chloride, Anhydrous, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Barium Chloride, Anhydrous, Thermo Scientific Chemicals

 
Have Questions?
Catalog NumberQuantity
612281000100 g
Catalog number 612281000
Price (USD)
185.65
Online Exclusive
206.00
Save 20.35 (10%)
Each
In stock
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)
185.65
Online Exclusive
206.00
Save 20.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS10361-37-2
IUPAC Namebarium(2+) dichloride
Molecular FormulaBaCl2
InChI KeyWDIHJSXYQDMJHN-UHFFFAOYSA-L
SMILES[Cl-].[Cl-].[Ba++]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay>=97 % (BaCl2)
Appearance (Color)White
Identificationpasses test
Appearance (Form)Granules
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
B0155989Certificate of AnalysisJun 11, 2024612281000
B0150335Certificate of AnalysisNov 14, 2023612281000
B0148061Certificate of AnalysisOct 20, 2023612281000
3 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection