Sea sand, Washed, Thermo Scientific Chemicals
Sea sand, Washed, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sea sand, Washed, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
500 g
25 kg
Packaging:
Plastic bottle
Catalog number 612350050
Price (USD)/ Each
123.65
Online exclusive
137.00 
Save 13.35 (10%)
-
Add to cart
Quantity:
5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
123.65
Online exclusive
137.00 
Save 13.35 (10%)
Add to cart
Sea sand, Washed, Thermo Scientific Chemicals
Catalog number612350050
Price (USD)/ Each
123.65
Online exclusive
137.00 
Save 13.35 (10%)
-
Add to cart
Chemical Identifiers
CAS14808-60-7
IUPAC Namesilanedione
Molecular FormulaO2Si
InChI KeyVYPSYNLAJGMNEJ-UHFFFAOYSA-N
SMILESO=[Si]=O
View more
SpecificationsSpecification SheetSpecification Sheet
Identificationpasses test
Appearance (Color)Light grey to tan
Appearance (Form)Crystals
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Filter aid
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only