Thermo Scientific Chemicals

Bromothymol Blue, 0.04% Aqueous Solution

Catalog number: 612405000
500 mL, EcoSafPak™, Each
Thermo Scientific Chemicals

Bromothymol Blue, 0.04% Aqueous Solution

Catalog number: 612405000
500 mL, EcoSafPak™, Each
Quantity
Packaging
Catalog number: 612405000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
34722-90-2
IUPAC Name
sodium 2-{[(1Z)-3-bromo-2-methyl-4-oxo-5-(propan-2-yl)cyclohexa-2,5-dien-1-ylidene][3-bromo-4-hydroxy-2-methyl-5-(propan-2-yl)phenyl]methyl}benzene-1-sulfonate
Molecular Formula
C27H27Br2NaO5S
InChI Key
NMKFVGALBGZKGW-FKWCIMQXSA-M
SMILES
[Na+].CC(C)C1=CC(\C(C2=CC=CC=C2S([O-])(=O)=O)=C2/C=C(C(C)C)C(=O)C(Br)=C2C)=C(C)C(Br)=C1O
Appearance (Color)
Clear dark green
Appearance (Form)
Liquid
Concentration
passes test
Additional info
visual transition interval: passes test
pH
6.5 to 7.1 (at 25°C)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text