Calcium Stearate, Thermo Scientific Chemicals
Calcium Stearate, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Calcium Stearate, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
3 kg
Catalog number 612430030
Price (USD)/ Each
-
Quantity:
3 kg
Request bulk or custom format
Chemical Identifiers
CAS66071-81-6
IUPAC Namecalcium dioctadecanoate
Molecular FormulaC36H70CaO4
InChI KeyCJZGTCYPCWQAJB-UHFFFAOYSA-L
SMILES[Ca++].CCCCCCCCCCCCCCCCCC([O-])=O.CCCCCCCCCCCCCCCCCC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Granular powder
Identificationpasses test
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only