Chemical Identifiers
CAS1340-02-9
IUPAC Nametetrasodium 2-[(4-{ethyl[(3-sulfonatophenyl)methyl]amino}phenyl)(4-{ethyl[(3-sulfonatophenyl)methyl]iminiumyl}cyclohexa-2,5-dien-1-ylidene)methyl]-5-hydroxybenzene-1-sulfonate bis(2-{2-[4-(dimethylamino)phenyl]diazen-1-yl}benzoate)
Molecular FormulaC67H62N8Na4O14S3
InChI KeyCNJVTYXLOYTWPD-UHFFFAOYSA-J
SMILES[Na+].[Na+].[Na+].[Na+].CN(C)C1=CC=C(C=C1)N=NC1=CC=CC=C1C([O-])=O.CN(C)C1=CC=C(C=C1)N=NC1=CC=CC=C1C([O-])=O.CCN(CC1=CC(=CC=C1)S([O-])(=O)=O)C1=CC=C(C=C1)C(=C1C=CC(C=C1)=[N+](CC)CC1=CC(=CC=C1)S([O-])(=O)=O)C1=C(C=C(O)C=C1)S([O-])(=O)=O
View more