Thermo Scientific Chemicals

Bromocresol Green, 0.04% Aqueous Solution

Catalog number: 612745000
500 mL, EcoSafPak™, Each
Thermo Scientific Chemicals

Bromocresol Green, 0.04% Aqueous Solution

Catalog number: 612745000
500 mL, EcoSafPak™, Each
Quantity
Packaging
Catalog number: 612745000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
62625-32-5
IUPAC Name
sodium 2,6-dibromo-4-[3-(3,5-dibromo-4-hydroxy-2-methylphenyl)-1,1-dioxo-3H-2,1λ⁶-benzoxathiol-3-yl]-3-methylbenzen-1-olate
Molecular Formula
C21H13Br4NaO5S
InChI Key
HEFSGAHJDGZCHA-UHFFFAOYNA-M
SMILES
[Na+].CC1=C(Br)C(O)=C(Br)C=C1C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC(Br)=C([O-])C(Br)=C1C
Appearance (Color)
Clear dark green
Visual transition interval
Passes test
Appearance (Form)
Liquid
Concentration
passes test
pH
4.3 to 4.9 (at 25°C)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text