Amphotericin B
Amphotericin B
Amphotericin B
Thermo Scientific Chemicals

Amphotericin B

CAS: 1397-89-3 | C47H73NO17 | 924.09 g/mol
Have Questions?
Catalog NumberQuantity
614122500250 mg
Catalog number 614122500
Price (USD)
119.65
Special offer
Online exclusive
140.00
Save 20.35 (15%)
Each
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (USD)
119.65
Special offer
Online exclusive
140.00
Save 20.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS1397-89-3
IUPAC Name(1R,3S,5R,6R,9R,11R,15S,16R,17R,18S,19Z,21Z,23Z,25Z,27Z,29Z,31Z,33R,35S,36R,37S)-33-{[(2R,3S,4S,5S,6R)-4-amino-3,5-dihydroxy-6-methyloxan-2-yl]oxy}-1,3,5,6,9,11,17,37-octahydroxy-15,16,18-trimethyl-13-oxo-14,39-dioxabicyclo[33.3.1]nonatriaconta-19,21,23,25,27,29,31-heptaene-36-carboxylic acid
Molecular FormulaC47H73NO17
InChI KeyAPKFDSVGJQXUKY-ZNVUZQDLSA-N
SMILESC[C@H]1O[C@@H](O[C@@H]2C[C@@H]3O[C@@](O)(C[C@H](O)[C@H]3C(O)=O)C[C@@H](O)C[C@@H](O)[C@H](O)CC[C@@H](O)C[C@@H](O)CC(=O)O[C@@H](C)[C@H](C)[C@H](O)[C@@H](C)\C=C/C=C\C=C/C=C\C=C/C=C\C=C/2)[C@@H](O)[C@@H](N)[C@@H]1O
View more
SpecificationsSpecification SheetSpecification Sheet
Identificationpasses test
Loss on drying=<5.0 % ()
Appearance (Color)Yellow
Appearance (Form)Powder
Additional infoamphotericin A: < 15%
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link