Nalgene™ Polypropylene, Barbed Bulkhead Fittings, 13 mm
Thermo Scientific™
Nalgene™ Polypropylene, Barbed Bulkhead Fittings, 13 mm
For Use With: Nalgene Bottle and Carboy Closures
Have Questions?
Catalog NumberQuantity
6149-0001Pack of 2
Catalog number 6149-0001
Price (USD)
378.00
Case of 24
Quantity:
Pack of 2
Price (USD)
378.00
Case of 24
Customize tubing sets as culture vessels for aseptic transfer of liquids with autoclavable Thermo Scientific™ Nalgene™ Barbed Polypropylene Bulkhead Fittings, suitable for use with most Nalgene bottle and carboy closures. Pre-drilled closures allow easy, safe in-house assembly.
Features:
  • Closures accept (two) 0.25″ fittings
  • Fittings, barbed at both ends, accept tubing for inside and outside container
Includes:
Instructions, drilling template and two each of polypropylene barbed fitting, acetyl nuts, silicone gaskets and and TPE port caps.
Specifications
Diameter (English) Inner Tubing0.5 in.
Diameter (Metric) Inner Tubing13 mm
For Use With (Equipment)Nalgene bottles and carboys
No. per Pack2
QuantityPack of 2
MaterialPolypropylene
Product TypeFitting
Unit SizeCase of 24