Nalgene™ Polypropylene, Barbed Bulkhead Fittings, 6.5 mm
Nalgene™ Polypropylene, Barbed Bulkhead Fittings, 6.5 mm
Thermo Scientific™
Nalgene™ Polypropylene, Barbed Bulkhead Fittings, 6.5 mm
For Use With: Nalgene Bottle and Carboy Closures
Have Questions?
Catalog NumberQuantity
6149-0002Case of 24
Catalog number 6149-0002
Price (USD)
315.00
Case of 24
Add to cart
Quantity:
Case of 24
Price (USD)
315.00
Case of 24
Add to cart
Customize tubing sets as culture vessels for aseptic transfer of liquids with autoclavable Thermo Scientific™ Nalgene™ Barbed Polypropylene Bulkhead Fittings, suitable for use with most Nalgene bottle and carboy closures. Pre-drilled 38mm-430 closures allow easy, safe in-house assembly.
Features:
  • 53B closures accept (two) 0.25″ fittings
  • Fittings, barbed at both ends, accept tubing for inside and outside container
Includes:
Instructions, drilling template and two each of polypropylene barbed fitting, acetyl nuts, silicone gaskets and and TPE port caps.
Specifications
Diameter (English) Inner Tubing0.25 in.
Diameter (Metric) Inner Tubing6.5 mm
For Use With (Equipment)Nalgene bottles and carboys
No. per Case24/Cs.
No. per Pack2 Pk.
QuantityCase of 24
MaterialPolypropylene
Product TypeFitting
Unit SizeCase of 24
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link