Specifications
Chemical Name or MaterialMethanol
Absorbance0.005 AU max. at 260nm, 0.005 AU max. at 280nm, 0.01 AU max. at 254nm, 0.1 AU max. at 230nm, 0.2 AU max. at 220nm, 0.4 AU max. at 214nm, 0.5 AU max. at 210nm
CAS67-56-1
Health Hazard 1Danger
Health Hazard 2GHS H Statement:
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes damage to organs.
Highly flammable liquid and vapor.
View more