1x1 image pixel for data collection
Nalgene™ Polypropylene Pinch Clamp
Nalgene™ Polypropylene Pinch Clamp
Thermo Scientific™
Nalgene™ Polypropylene Pinch Clamp
From completely open to restricted flow to completely closed—experience precise, one-hand regulation of fluid flow with the Thermo Scientific™ Nalgene™ Polypropylene Pinch Clamp.
Have Questions?
Catalog NumberQuantity
6165-0002Case of 72
Catalog number 6165-0002
Price (USD)
207.00
Case of 72
In stock
Add to cart
Quantity:
Case of 72
Recurring order eligible. Learn more »
Price (USD)
207.00
Case of 72
Add to cart
Ask our AI about this Product
From completely open to restricted flow to completely closed—experience precise, one-hand regulation of fluid flow with the Thermo Scientific™ Nalgene™ Polypropylene Pinch Clamp. This plastic clamp will not rust, corrode or deform. Molded in one piece with no sharp edges, it accommodates a wide range of tubing.
Specifications
DescriptionPinch clamp; Fits tubing with O.D.: 1/4 to 7/16 in.
For Use With (Equipment)Fits tubing with O.D.: 1/4 to 7/16 in.
QuantityCase of 72
MaterialPolypropylene
TypePinch Clamp
Unit SizeCase of 72
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Customers who viewed this item also viewed
Documents & Downloads
Certificates
0 results displayed, search above for a specific certificate
Scientific Resources
Share catalog number, name or link
1x1 image pixel for data collection