Informational:Discover the future of Gibco Media selection with our upcoming updated experience. View details >
Gibco™

RPMI 1640 Medium, GlutaMAX™ Supplement

Catalog number: 61870143
Have Questions?
Gibco™

RPMI 1640 Medium, GlutaMAX™ Supplement

Catalog number: 61870143
Have Questions?
Catalog Number
61870143
Unit Size
5 L
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
10 L
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
618701435 L
-
6187015010 L
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
RPMI 1640 Medium (Roswell Park Memorial Institute 1640 Medium) was originally developed to culture human leukemic cells in suspension and as a monolayer. RPMI 1640 medium has since been found suitable for a variety of mammalian cells, including HeLa, Jurkat, MCF-7, PC12, PBMC, astrocytes, and carcinomas. We offer a variety of RPMI 1640 modifications for a range of cell culture applications. Find the right formulation using the media selector tool.

This RPMI 1640 is modified as follows:
WithWithout
• GlutaMAX™• HEPES
• Phenol Red

The complete formulation is available.

Using RPMI 1640 Medium
RPMI 1640 medium is unique from other media because it contains the reducing agent glutathione and high concentrations of vitamins. RPMI 1640 medium contains biotin, vitamin B12, and PABA, which are not found in Eagle's Minimal Essential Medium or Dulbecco's Modified Eagle Medium. In addition, the vitamins inositol and choline are present in very high concentrations. RPMI 1640 medium with GlutaMAX™ supplement minimizes toxic ammonia build-up and improves cell viability and growth in an easy-to-use format. RPMI 1640 medium contains no proteins, lipids, or growth factors. Therefore, RPMI 1640 medium requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). RPMI 1640 medium uses a sodium bicarbonate buffer system (2.0 g/L) and therefore requires a 5–10% CO2 environment to maintain physiological pH.

cGMP manufacturing and quality system
RPMI 1640 is manufactured at a cGMP-compliant facility located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards. For supply chain continuity, we offer an identical RPMI 1640 product made in our Scotland facility (61870-010). This facility is also registered with the FDA as a medical device manufacturer and certified to the ISO 13485 standard.
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.

Specifications

Cell Line
HeLa, Jurkat, MCF-7, PC-12, PBMC, astrocytes, and carcinomas
Cell Type
Leukemic Cells
Classification
Animal Origin-free
Concentration
1 X
Form
Liquid
Product Type
RPMI 1640 Medium (Roswell Park Memorial Institute 1640 Medium)
Serum Level
Standard Serum Supplementation
Shelf Life
12 Months From Date of Manufacture
Sterility
Sterile-filtered
With Additives
GlutaMAX, Phenol Red
Without Additives
No HEPES, No Sodium Pyruvate
Product Line
Gibco™, GlutaMAX™, SynerGy™
Quantity
5 L
Shipping Condition
Room Temperature

Contents & Storage

Storage conditions: 2-8° C. Protect from light
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text